Learn More
anti-Digoxin, Clone: 26G10, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus Biologicals NBP178619
Catalog No. NBP178619
Add to cart
Description
Description
Digoxin Monoclonal antibody specifically detects Digoxin in Non-species specific samples. It is validated for Western Blot, ELISA, Immunoprecipitation, Radioimmunoassay, Chemiluminescence Immunoassay.
Specifications
Specifications
Digoxin
26G10
Unconjugated
10mM Potassium Phosphate, 2.5mM Potassium Chloride, 135mM NaCl and 1.0 mg/ml BSA with 0.05% Sodium Azide
Lanoxin
Digoxin-BSA
Ion exchange chromatography
RUO
Primary
Chemical.
Chemiluminescence Immunoassay, ELISA, Immunoprecipitation, Radioimmune Assays, Western Blot
1 mg/mL
Western Blot 1 ug/ml, ELISA 1:100 - 1:2000, Immunoprecipitation 1:10 - 1:500, Radioimmunoassay 1:100 - 1:2000, Chemiluminescence Immunoassay
Affinity Purified
Mouse
IgG1
100ug
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Monoclonal
Other