Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DNA Ligase I Antibody (10H5), Novus Biologicals™
Mouse Monoclonal Antibody has been used in 23 publications
Supplier: Novus Biologicals NB100119
Description
DNA Ligase I Monoclonal specifically detects DNA Ligase I in Human, Mouse, Bacteria, Chicken, Fish, Hamster, Rabbit samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, In vitro assay, Radioimmunoassay.Specifications
DNA Ligase I | |
Monoclonal | |
1mg/mL | |
Western Blot, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, In vitro assay, Radioimmunoassay | |
LIG1 | |
Full-length recombinant human DNA Ligase I protein | |
Protein G purified | |
RUO | |
Primary | |
This is specific for DNA ligase 1. | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
IgG1 |
Western Blot, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin), In vitro Assay, Radioimmune Assays (RIA) | |
10H5 | |
Unconjugated | |
DNA ligase 1, DNA ligase I, EC 6.5.1.1, ligase I, DNA, ATP-dependent, MGC117397, MGC130025, Polydeoxyribonucleotide synthase [ATP] 1, polydeoxyribonucleotide synthase ATP 1 | |
Mouse | |
125 kDa | |
100 μL | |
Apoptosis, Base Excision Repair, Cancer, DNA Repair, Nucleotide Excision Repair, Tumor Suppressors | |
3978 | |
Human, Mouse | |
Purified |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?
Provide Content Correction