Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DNA Ligase III Antibody (1F3), Novus Biologicals™
Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP141190
Description
DNA Ligase III Monoclonal specifically detects DNA Ligase III in Human, Mouse, Chicken samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Proximity Ligation Assay, Block/Neutralize.Specifications
DNA Ligase III | |
Monoclonal | |
1.0mg/mL | |
Western Blot 1:500-1:3000, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Proximity Ligation Assay, Block/Neutralize | |
LIG3 | |
Recombinant full-length human DNA Ligase III-b. | |
Immunogen affinity purified | |
RUO | |
Primary | |
This antibody will recognize human DNA ligase III. | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
IgG1 |
Western Blot, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Blocking Assay, Proximity Ligation Assay | |
1F3 | |
Unconjugated | |
DNA ligase 3, DNA ligase III, EC 6.5.1.1, LIG2, ligase II, DNA, ATP-dependent, ligase III, DNA, ATP-dependent, Polydeoxyribonucleotide synthase [ATP] 3 | |
Mouse | |
100 kDa | |
100 μL | |
Base Excision Repair, Cancer, DNA Repair | |
3980 | |
Human, Mouse, Chicken | |
Purified |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?
Provide Content Correction