Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-Mouse DOG1/TMEM16A, Clone: DOG1.1, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP149559

Catalog No. NBP149559


Add to cart

Description

Description

DOG1/TMEM16A Monoclonal antibody specifically detects DOG1/TMEM16A in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.
Specifications

Specifications

DOG1/TMEM16A
DOG1.1
Unconjugated
Tissue culture supernatant, containing less than 0.1% sodium azide with Sodium Azide
Affinity Purified
ANO1
A synthetic peptide of human DOG1 protein.
Tissue culture supernatant
RUO
Primary
55107
Human
Immunohistochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin)
Predilute
Immunohistochemistry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen
Q5XXA6
ANO1, anoctamin 1, DOG1, ORAOV2, TAOS2, TMEM16A
Mouse
IgG
6 ml
Store at 4°C.
Monoclonal
DOG1/TMEM16A is expressed ubiquitously in gastrointestinal stromal tumors irrespective of c-kit or PDGFR alpha mutation status. Reactivity for DOG1/TMEM16A may aid in the diagnosis of gastrointestinal stromal tumors, including PDGFRA mutants that fail to express c-kit antigen, and lead to appropriate treatment with imatinib mesylate, an inhibitor of the c-kit tyrosine kinase.
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only