Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Doublecortin Mouse anti-Human, Mouse, Rat, Porcine, Bovine, Equine, DyLight 405, Clone: 30, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP192684V

Catalog No. NBP192684V


Add to cart

Description

Description

Doublecortin Monoclonal antibody specifically detects Doublecortin in Human, Mouse, Rat, Porcine, Bovine, Equine samples. It is validated for Western Blot, Immunocytochemistry, Immunofluorescence.
Specifications

Specifications

Doublecortin
30
DyLight 405
50mM Sodium Borate with 0.05% Sodium Azide
Purified
DCX
Recombinant full length human Lis-A isoform of Doublecortin purified from E. coli. [UniProt# O43602]
49.3 kDa
0.1 ml
Neuroscience, Phospho Specific, Signal Transduction
Primary
1641
Bovine, Equine, Human, Mouse, Porcine, Rat
Immunocytochemistry, Immunofluorescence, Western Blot
1mg/mL
Western Blot, Immunocytochemistry/Immunofluorescence
O43602
doublecortin, Doublin, FLJ51296, Lissencephalin-X, Lis-X, X-linked (doublecortin)
Mouse
IgG2a
Protein G purified
RUO
Store at 4C in the dark.
Monoclonal
Human, Mouse, Rat, Cow, Pig and other Mammals.
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only