Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Doublecortin Antibody (3E1), DyLight 488, Novus Biologicals™
Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP192684G
Description
Doublecortin Monoclonal specifically detects Doublecortin in Human, Mouse, Rat, Porcine, Bovine, Equine samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.Specifications
Doublecortin | |
Monoclonal | |
DyLight 488 | |
O43602 | |
DCX | |
Recombinant full length human Lis-A isoform of Doublecortin purified from E. coli. [UniProt# O43602] | |
Protein G purified | |
RUO | |
Primary | |
Human, Mouse, Rat, Cow, Pig and other Mammals. | |
Store at 4C in the dark. | |
IgG2a |
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
30 | |
Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence | |
doublecortin, Doublin, FLJ51296, Lissencephalin-X, Lis-X, X-linked (doublecortin) | |
Mouse | |
49.3 kDa | |
0.1 mL | |
Neuroscience, Phospho Specific, Signal Transduction | |
1641 | |
Human, Mouse, Rat, Porcine, Bovine, Equine | |
Purified |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only