Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-Dtk/TYRO3, Clone: 6D6F10, Novus Biologicals™

Manufacturer:  Novus Biologicals NBP147376

Catalog No. NBP147376

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Specifications

Specifications

Dtk/TYRO3
6D6F10
ELISA 1:10000, Immunocytochemistry/Immunofluorescence 1:50 - 1:200
Ascites
TYRO3
Purified recombinant fragment of TYRO3(aa138-321) expressed in E. Coli.
Unpurified
RUO
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Monoclonal
Human
ELISA, Immunocytochemistry, Immunofluorescence
Unconjugated
Ascites with 0.03% Sodium Azide
Brt, BYK, DTK, EC 2.7.10, EC 2.7.10.1, FLJ16467, RSE, Sky, Tif, TYRO3 protein tyrosine kinase, Tyrosine-protein kinase byk, Tyrosine-protein kinase DTK, tyrosine-protein kinase receptor TYRO3, Tyrosine-protein kinase RSE, Tyrosine-protein kinase SKY
Mouse
IgG1
0.1 ml
Cancer, Neuroscience, Neurotransmission, Protein Kinase, Tyrosine Kinases
Primary
7301
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only