Learn More
anti-DUX4, Alexa Fluor 488, Clone: P4H2, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus Biologicals NBP149552AF488
Catalog No. NBP149552X
Add to cart
Description
Description
DUX4 Monoclonal antibody specifically detects DUX4 in Human samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.
Specifications
Specifications
DUX4
P4H2
Western Blot, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen
Q9UBX2
double homeobox 4, double homeobox protein 10, double homeobox protein 4, double homeobox protein 4/10, double homeobox protein DUX10, DUX10
Mouse
IgG1
Protein G purified
RUO
Store at 4C in the dark.
Monoclonal
This antibody is specific for DUX4 and does not recognize DUX4c.
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot
Alexa Fluor 488
50mM Sodium Borate with 0.05% Sodium Azide
Purified
DUX4
Synthetic peptide corresponding to the C-terminus of human DUX4. [Swiss-Prot: Q9UBX2]
52 kDa
0.1 ml
Neuroscience, Transcription Factors and Regulators
Primary
100288687
Human