Learn More
Ebola virus Mouse anti-Ebola Virus, Virus, Clone: FE18, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus Biologicals NB1107381
Catalog No. NB1107381
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
Ebola virus Monoclonal antibody specifically detects Ebola virus in Ebola Virus, Virus samples. It is validated for Western Blot, ELISA.
Specifications
Specifications
Ebola virus
FE18
Western Blot 1:100-1:2000, ELISA 1:100-1:2000
Purified
Formalin inactivated Ebola virus (strain Zaire).
Protein G purified
RUO
Primary
Ebola virus (FE18)
Western Blot, ELISA
Unconjugated
PBS (pH 7.2) with 0.09% Sodium Azide
Mouse
IgG2a
0.2 mg
Store at 4C. Do not freeze.
Monoclonal
Ebola Virus, Virus