Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-EGF, Clone: 4E11, Novus Biologicals™

Manufacturer:  Novus Biologicals NBP147473

Catalog No. NBP147473

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Specifications

Specifications

EGF
400000000000
Western Blot 1:500 - 1:2000, ELISA 1:10000, Immunocytochemistry/Immunofluorescence 1:200 - 1:1000
P01133
beta-urogastrone, epidermal growth factor, epidermal growth factor (beta-urogastrone), HOMG4, pro-epidermal growth factor, URG
Mouse
IgG1
Unpurified
RUO
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Monoclonal
Human
Western Blot, ELISA, Immunocytochemistry, Immunofluorescence
Unconjugated
Ascites with 0.03% Sodium Azide
Ascites
EGF
Purified recombinant fragment of human EGF expressed in E. Coli.
134 kDa
0.1 ml
Angiogenesis, Cancer, Cell Cycle and Replication, Cytokine Research, Immunology, Virology Bacteria and Parasites
Primary
1950
SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only