Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
EGLN3/PHD3 Antibody (EG188e/d5), DyLight 650, Novus Biologicals™
Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP130440C
Description
EGLN3/PHD3 Monoclonal specifically detects EGLN3/PHD3 in Human, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
EGLN3/PHD3 | |
Monoclonal | |
DyLight 650 | |
Q9H6Z9 | |
EGLN3 | |
Partial recombinant human PHD3/HIF Prolyl Hydroxylase 3 (UniProt# Q9H6Z9) | |
Protein G purified | |
RUO | |
Primary | |
Human, monkey and rat. Immunogen sequence has 98% identity to bovine and mouse. | |
Store at 4C in the dark. | |
IgG1 |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
EG188e/d5 | |
Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin | |
EC 1.14.11, EC 1.14.11.-, EGL nine (C.elegans) homolog 3, egl nine homolog 3, egl nine homolog 3 (C. elegans), egl nine-like protein 3 isoform, FLJ21620, HIF prolyl hydroxylase 3, HIF-PH3, HIFPH3 MGC125998, HIF-prolyl hydroxylase 3, HPH-1, HPH-3, Hypoxia-inducible factor prolyl hydroxylase 3, MGC125999, pdh3, PHD3, Prolyl hydroxylase domain-containing protein 3 | |
Mouse | |
27 kDa | |
0.2 mL | |
Apoptosis, Cancer, Chromatin Research, Hypoxia | |
112399 | |
Human, Rat | |
Purified |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?
Provide Content Correction