Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-eIF5A, Alexa Fluor 647, Clone: 2E9, Novus Biologicals™

Manufacturer:  Novus Biologicals NBP148352AF647

Catalog No. NBP148352Y

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Specifications

Specifications

eIF5A
2000000000
Western Blot, ELISA, Immunocytochemistry/Immunofluorescence
NP_001961
eIF-4D, eIF-5A, eIF-5A1, eIF-5A-1, EIF5A1, eIF5AI, eukaryotic initiation factor 5A, Eukaryotic initiation factor 5A isoform 1, eukaryotic translation initiation factor 5A, eukaryotic translation initiation factor 5A-1, MGC104255, MGC99547, Rev-binding factor
Mouse
IgG1
0.1 ml
Store at 4C in the dark.
Monoclonal
Human EIF5A (1-154aa)
ELISA, Immunocytochemistry, Immunofluorescence, Western Blot
Alexa Fluor 647
50mM Sodium Borate with 0.05% Sodium Azide
Purified
EIF5A
Recombinant human EIF5A (1-154aa) purified from E. coli.
Protein A or G purified
RUO
Primary
1984
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only