Learn More
Mouse EphB2 Biotinylated Antibody, R&D Systems™
Goat Polyclonal Antibody
Supplier:  R&D Systems BAF467
Catalog No. BAF467
Add to Cart
This item is not returnable. View return policy
Description
Description
EphB2 Polyclonal specifically detects EphB2 in Mouse samples. It is validated for Western Blot.
Specifications
Specifications
EphB2
Polyclonal
Biotin
P54763
EPHB2
Mouse myeloma cell line NS0-derived recombinant mouse EphB2 Met1-Lys548 Accession # P54763
50 μg
Primary
Detects mouse EphB2 in Western blots. In Western blots, approximately 5% cross-reactivity with recombinant mouse (rm) EphB6, recombinant rat EphB1, and rmEphB3 is observed.
Mouse
IgG
Western Blot
LYOPH
Western Blot 0.1 ug/mL
CAPB, Cek5, Drt, DRTEphB2, EC 2.7.10, EC 2.7.10.1, EK5, elk-related tyrosine kinase, EPH receptor B2, eph tyrosine kinase 3, EPH-like kinase 5, ephrin type-B receptor 2, EPHT3MGC87492, EPTH3, Erk, ERKHek5, HEK5, Nuk, PCBC, protein-tyrosine kinase HEK5, Qek2, Renal carcinoma antigen NY-REN-47, Sek3, Tyro5, Tyrosine-protein kinase receptor EPH-3, Tyrosine-protein kinase TYRO5
Goat
Affinity Purified
RUO
2048
Reconstitute at 0.2 mg/mL in sterile PBS.
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Product Suggestions
Product Suggestions