Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-ER beta/NR3A2, Clone: 4D2, Novus Biologicals™

Manufacturer:  Novus Biologicals NBP141193

Catalog No. NBP141193

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Specifications

Specifications

ER beta/NR3A2
4D2
Unconjugated
PBS (pH 7.4) with No Preservative
Erb, ER-beta, ESR-BETA, ESTRB, estrogen receptor 2 (ER beta), estrogen receptor beta, estrogen receptor beta 4, NR3A2ESRB, Nuclear receptor subfamily 3 group A member 2
Mouse
IgG2b
100ul
Breast Cancer, Cancer, GPCR
Primary
2100
Human
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
1.0mg/mL
Western Blot 1-5 ug/ml, Immunohistochemistry 10 - 1:500, Immunohistochemistry-Paraffin 1:10 - 1:500
Purified
ESR2
Amino acids 1-153 of human ER expressed in E. coli.
Immunogen affinity purified
RUO
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Monoclonal
This antibody will recognize human ER beta.
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only