Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-ERK2, Clone: 3F8, Novus Biologicals™

Manufacturer:  Novus Biologicals NBP142563

Catalog No. NBP142563

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Specifications

Specifications

ERK2
3F8
Western Blot 1:500-1:2000, Flow Cytometry 1:200-1:400, ELISA 1:10000, Immunohistochemistry-Paraffin
Ascites
MAPK1
Purified recombinant fragment of human MAPK1 expressed in E. Coli.
42 kDa
0.1 ml
Angiogenesis, Apoptosis, Breast Cancer, Cancer, Diabetes Research, Hypoxia, Protein Kinase, Signal Transduction, Tumor Suppressors
Primary
5594
Western Blot, Flow Cytometry, ELISA, Immunohistochemistry (Paraffin)
Unconjugated
Ascites with 0.03% Sodium Azide
EC 2.7.11, EC 2.7.11.24, ERK, ERK-2, ERK2MAP kinase isoform p42, ERT1, Extracellular signal-regulated kinase 2, MAP kinase 1, MAP kinase 2, MAPK 1, MAPK2, mitogen-activated protein kinase 1, Mitogen-activated protein kinase 2, p38, p40, p41, p41mapk, p42-MAPK, P42MAPK, PRKM1MAPK 2, PRKM2, protein tyrosine kinase ERK2
Mouse
IgG2b
Unpurified
RUO
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Monoclonal
Human, Mouse
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only