Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ERK2 Mouse anti-Human, Mouse, Rat, Canine, Monkey, Clone: OTI6E5, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP147842

Catalog No. NBP147842


Add to cart

Description

Description

ERK2 Monoclonal specifically detects ERK2 in Human, Mouse, Rat, Canine, Monkey samples. It is validated for Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Paraffin.
Specifications

Specifications

ERK2
OTI6E5
Unconjugated
PBS (pH 7.3), 1.0% BSA and 50% Glycerol with 0.02% Sodium Azide
EC 2.7.11, EC 2.7.11.24, ERK, ERK-2, ERK2MAP kinase isoform p42, ERT1, Extracellular signal-regulated kinase 2, MAP kinase 1, MAP kinase 2, MAPK 1, MAPK2, mitogen-activated protein kinase 1, Mitogen-activated protein kinase 2, p38, p40, p41, p41mapk, p42-MAPK, P42MAPK, PRKM1MAPK 2, PRKM2, protein tyrosine kinase ERK2
Mouse
IgG1
Immunogen affinity purified
RUO
Store at -20°C. Avoid freeze-thaw cycles.
Monoclonal
This antibody is specific for Homo sapiens mitogen-activated protein kinase 1 (MAPK1), transcript variant 2.
Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin)
1 mg/mL
Western Blot 1:2000, Immunohistochemistry 1:50, Immunoprecipitation 2-4ug/mg, Immunohistochemistry-Paraffin 1:50
Affinity Purified
MAPK1
Full length human recombinant protein of human MAPK1(NP_620407) produced in HEK293T cell.
41.4 kDa
0.1 ml
Angiogenesis, Apoptosis, Breast Cancer, Cancer, Diabetes Research, Hypoxia, Protein Kinase, Signal Transduction, Tumor Suppressors
Primary
5594
Human, Mouse, Rat, Canine, Monkey
SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only