Learn More
EWSR1 Mouse anti-Human, Mouse, Rat, Canine, Equine, DyLight 350, Clone: 5H7, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP192686UV
Catalog No. NBP192686UV
Add to cart
Description
Description
EWSR1 Monoclonal antibody specifically detects EWSR1 in Human, Mouse, Rat, Canine, Equine samples. It is validated for Western Blot, Immunocytochemistry, Immunofluorescence.
Specifications
Specifications
EWSR1
5H7
Western Blot, Immunocytochemistry/Immunofluorescence
Q01844
bK984G1.4, Ewing sarcoma breakpoint region 1, Ewing sarcoma breakpoint region 1 protein, Ewings sarcoma EWS-Fli1 (type 1) oncogene, EWS oncogene, EWSRNA-binding protein EWS
Mouse
IgG2b Kappa
Protein G purified
RUO
Store at 4C in the dark.
Monoclonal
Canine, Equine, Human, Mouse, Rat
Immunocytochemistry, Immunofluorescence, Western Blot
DyLight 350
50mM Sodium Borate with 0.05% Sodium Azide
Purified
EWSR1
Full length recombinant human EWSR1 expressed in and purified from E. coli. [UniProt# Q01844]
85 kDa
0.1 ml
Cancer, Cellular Markers, Neuroscience, Tumor Suppressors
Primary
2130