Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-FABP3/H-FABP, Clone: 67D3, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP142507

Catalog No. NBP142507


Add to cart

Description

Description

FABP3/H-FABP Monoclonal antibody specifically detects FABP3/H-FABP in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.
Specifications

Specifications

FABP3/H-FABP
67D3
Unconjugated
PBS containing 0.1% BSA with 0.02% Sodium Azide
FABP11, fatty acid binding protein 11, fatty acid binding protein 3, muscle and heart (mammary-derived growthinhibitor), Fatty acid-binding protein 3, Fatty acid-binding protein 3, muscle, fatty acid-binding protein, heart, Heart-type fatty acid-binding protein, H-FABPM-FABP, Mammary-derived growth inhibitor, MDGI, Muscle fatty acid-binding protein, O-FABP
Mouse
IgG1
0.2 ml
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Monoclonal
This monoclonal antibody (subclass IgG1) binds to human heart Fatty Acid Bindings Protein. It shows no binding to human intestinal or liver FABP. The antibody stains heart muscle cells and striated skeletal muscle cells in immunohistochemistry. The antibody can be used to detect ischemia areas of human heart. The antibody is cross reactive with rat and mouse H-FABP.
ELISA, Immunohistochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
0.1 mg/mL
Western Blot 1:50-1:100, ELISA 1:100-1:2000, Immunohistochemistry 1:10-1:500, Immunoprecipitation 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:500, Immunohistochemistry-Frozen 1:10-1:500
Purified
FABP3
The antibody reacts with the H-FABP antigen.
Protein A or G purified
RUO
Primary
2170
Human, Mouse, Rat
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only