Learn More
anti-FABP4/A-FABP, Clone: 1105CT1.1.1, Novus Biologicals™
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP232295
Catalog No. NBP232295
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Specifications
Specifications
FABP4/A-FABP
1105CT1.1.1
Unconjugated
PBS with 0.09% Sodium Azide
Purified
FABP4
Purified His-tagged FABP4 protein was used to produced this monoclonal antibody.
14.719 kDa
400 ul
Cancer, Phospho Specific, Stem Cell Markers
Primary
2167
Western Blot, Immunohistochemistry
0.48 mg/mL
Western Blot 1:1000, Immunohistochemistry 1:10-1:500
P15090, P15090
Adipocyte lipid-binding protein, Adipocyte-type fatty acid-binding protein, A-FABPAFABP, ALBP, aP2, fatty acid binding protein 4, adipocyte, Fatty acid-binding protein 4, fatty acid-binding protein, adipocyte
Mouse
IgG1
Protein G purified
RUO
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Monoclonal
Human, Mouse