Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-FABP4/A-FABP, Clone: 3F4, Novus Biologicals™

Manufacturer:  Novus Biologicals NBP123021

Catalog No. NBP123021

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Specifications

Specifications

FABP4/A-FABP
3F4
Unconjugated
PBS (pH 7.4) and 10% Glycerol with 0.02% Sodium Azide
Purified
FABP4
Recombinant human FABP4 (1-132aa) purified from E. coli
Protein G purified
RUO
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Monoclonal
Anti-human FABP4 mAb, clone 3F4, is derived from hybridization of mouse F0 myeloma cells with spleen cells from BALB/c mice immunized with a recombinant human FABP4 protein.
Western Blot, ELISA
1.0 mg/mL
Western Blot 1:500-1:2000, ELISA 1:100-1:2000
NP_001433
Adipocyte lipid-binding protein, Adipocyte-type fatty acid-binding protein, A-FABPAFABP, ALBP, aP2, fatty acid binding protein 4, adipocyte, Fatty acid-binding protein 4, fatty acid-binding protein, adipocyte
Mouse
IgG1 Kappa
0.1 mL
Cancer, Stem Cell Markers
Primary
2167
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only