Learn More
anti-FAK, Clone: 1297CT261.163.145, Novus Biologicals™
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP232307
Catalog No. NBP232307
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Specifications
Specifications
FAK
1297CT261.163.145
Unconjugated
PBS with 0.09% Sodium Azide
Purified
PTK2
This PTK2 antibody is generated from a mice immunized with a recombinant protein between 1-128 amino acids from the N-terminal region of human PTK2.
119.233 kDa
400 ul
Angiogenesis, Cancer, Cellular Markers, Extracellular Matrix, Phospho Specific, Protein Kinase, Signal Transduction, Tyrosine Kinases
Primary
5747
Western Blot
0.5 mg/mL
Western Blot 1:1000
Q05397
EC 2.7.10, FADK 1, FADKFAK-related non-kinase polypeptide, FAK1FRNK, FAKEC 2.7.10.2, focal adhesion kinase 1, pp125FAK, Protein-tyrosine kinase 2, PTK2 protein tyrosine kinase 2
Mouse
IgG1
Protein G purified
RUO
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Monoclonal
Human