Learn More
anti-FDC, DyLight 680, Clone: CNA.42, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus Biologicals NBP143118FR
Catalog No. NBP143118FR
Add to cart
Description
Description
FDC Monoclonal antibody specifically detects FDC in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.
Specifications
Specifications
FDC
CNA.42
Western Blot, Flow Cytometry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen
Affinity Purified
CMD1B
The immunogen for this antibody was follicular dendritic cells.
Protein A or G purified
RUO
Primary
1218
Flow Cytometry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot
DyLight 680
50mM Sodium Borate with 0.05% Sodium Azide
cardiomyopathy, dilated 1B (autosomal dominant), CMPD1, FDC
Mouse
IgM
0.1 ml
Store at 4C in the dark.
Monoclonal
Human