Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-FDC, PerCP, Clone: CNA.42, Novus Biologicals™

Manufacturer:  Novus Biologicals NBP143118PCP

Catalog No. NBP143118P

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Specifications

Specifications

FDC
CNA.42
Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen
Affinity Purified
CMD1B
The immunogen for this antibody was follicular dendritic cells.
Protein A or G purified
RUO
Primary
1218
Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot
PerCP
PBS with 0.05% Sodium Azide
cardiomyopathy, dilated 1B (autosomal dominant), CMPD1, FDC
Mouse
IgM
0.1 ml
Store at 4C in the dark.
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only