Learn More
anti-Ferritin Light Chain, Clone: 521, Novus Biologicals™
Manufacturer:  Novus Biologicals NBP178567
Catalog No. NBP178567
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Specifications
Specifications
Ferritin Light Chain
521
Unconjugated
10mM Potassium Phosphate, 2.5mM Potassium Chloride, 135mM NaCl and 1.0 mg/ml BSA with 0.05% Sodium Azide
Purified
FTL
Human Liver Ferritin
Protein A purified
RUO
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Monoclonal
Ferritin Light Chain
ELISA, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Radioimmune Assays
1 mg/mL
ELISA 1 ug/ml, Immunocytochemistry/Immunofluorescence 1:50 - 1:200, Immunoprecipitation 1:10 - 1:500, Radioimmunoassay 1:100 - 1:2000
P02792
Ferritin L subunit, ferritin L-chain, ferritin light chain, ferritin light polypeptide-like 3, ferritin, light polypeptide, MGC71996, NBIA3
Mouse
IgG1
100ug
Cancer
Primary
2512
Human