Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-Fes, Clone: 2E3G3, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP128880

Catalog No. NBP128880


Add to cart

Description

Description

Fes Monoclonal antibody specifically detects Fes in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.
Specifications

Specifications

Fes
2E3G3
Western Blot 1:100-1:2000, ELISA 1:100-1:2000, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:500
Ascites
FES
Purified recombinant fragment of FES (AA:613-822)expressed in E. Coli.
Unpurified
RUO
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Monoclonal
Specific for FES.
ELISA, Immunohistochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot
Unconjugated
Ascites with 0.03% Sodium Azide
EC 2.7.10, EC 2.7.10.2, feline sarcoma oncogene, feline sarcoma virus, Proto-oncogene c-Fes, Proto-oncogene c-Fps, proto-oncogene tyrosine-protein kinase Fes/Fps, tyrosine-protein kinase Fes/Fps
Mouse
IgG1
0.1 ml
Protein Kinase
Primary
2242
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only