Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fibulin 2 Mouse anti-Human, Clone: 7H4F6, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP128889

Catalog No. NBP128889


Add to cart

Description

Description

Fibulin 2 Monoclonal antibody specifically detects Fibulin 2 in Human samples. It is validated for Western Blot, ELISA.
Specifications

Specifications

Fibulin 2
7H4F6
Western Blot 1:500-1:2000, ELISA 1:10000
Ascites
FBLN2
Purified recombinant fragment of FBLN2(aa180-440) expressed in E. Coli.
Unpurified
RUO
Primary
2199
Human, Monkey
ELISA, Western Blot
Unconjugated
Ascites with 0.03% Sodium Azide
FIBL-2, fibulin 2, fibulin-2
Mouse
IgG1
0.1 ml
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Monoclonal
Specific for FBLN2.
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only