Learn More
anti-FIH-1/HIF-1AN, DyLight 488, Clone: 162c, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus Biologicals NBP130333G
Catalog No. NBP130333G
Add to cart
Description
Description
FIH-1/HIF-1AN Monoclonal antibody specifically detects FIH-1/HIF-1AN in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications
Specifications
FIH-1/HIF-1AN
162c
Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin
Q9NWT6
DKFZp762F1811, EC 1.14.11.16, factor inhibiting HIF1, Factor inhibiting HIF-1, FIH1FIH-1, FLJ20615, FLJ22027, hypoxia inducible factor 1, alpha subunit inhibitor, hypoxia-inducible factor 1-alpha inhibitor, Hypoxia-inducible factor asparagine hydroxylase, peptide-aspartate beta-dioxygenase
Mouse
IgG1
Protein G purified
RUO
Store at 4C in the dark.
Monoclonal
Human
Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot
DyLight 488
50mM Sodium Borate with 0.05% Sodium Azide
Purified
HIF1AN
Full length human Factor Inhibiting HIF-1 protein [Swiss-Prot# Q9NWT6]
40 kDa
0.1 ml
Angiogenesis, Cancer, Chromatin Research, Hypoxia, Transcription Factors and Regulators
Primary
55662