Learn More
anti-Filamin A, Clone: 1273CT424.104.153, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP232299
Catalog No. NBP232299
Add to cart
Description
Description
Filamin A Monoclonal antibody specifically detects Filamin A in Human, Rat samples. It is validated for Western Blot.
Specifications
Specifications
Filamin A
1273CT424.104.153
Unconjugated
PBS with 0.09% Sodium Azide
Purified
FLNA
Purified His-tagged FLNA protein was used to produced this monoclonal antibody.
280.739 kDa
400 ul
Neuroscience, Phospho Specific
Primary
2316
Western Blot
0.44 mg/mL
Western Blot 1:1000
P21333
ABP-280DKFZp434P031, ABPX, actin binding protein 280, Actin-binding protein 280, Alpha-filamin, CVD1, EC 2.1.1.43, EC 6.3.4.4, Endothelial actin-binding protein, filamin A, alpha, filamin A, alpha (actin binding protein 280), Filamin-1, filamin-A, FLN, FLN1OPD, FLN-A, FMD, MNS, NHBP, Non-muscle filamin, OPD1, OPD2, XLVD, XMVD
Mouse
IgG1
Protein G purified
RUO
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Monoclonal
Human, Rat