Learn More
anti-Flt-3/Flk-2/CD135, DyLight 755, Clone: BV10A4, Novus Biologicals™
Manufacturer:  Novus Biologicals NBP142792IR
Catalog No. NBP142792IR
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Specifications
Specifications
Flt-3/Flk-2/CD135
BV10A4
Flow Cytometry
Purified
FLT3
BV-173 leukemic cell line
Protein A purified
RUO
Store at 4C in the dark.
Monoclonal
The mouse monoclonal antibody BV10A4 (BV10) reacts with CD135 (FLT3, FLK2, STK-1), a 130-160 kDa type III receptor tyrosine kinase that is involved in early steps of hematopoiesis.
Flow Cytometry
DyLight 755
50mM Sodium Borate with 0.05% Sodium Azide
CD135, CD135 antigen, EC 2.7.10, fetal liver kinase 2, FL cytokine receptor, FLK2, FLT-3, FLT3 receptor tyrosine kinase, Fms-like tyrosine kinase 3, fms-related tyrosine kinase 3, growth factor receptor tyrosine kinase type III, Stem cell tyrosine kinase 1, STK-1, STK1EC 2.7.10.1, Tyrosine-protein kinase receptor FLT3
Mouse
IgG1
0.1 ml
Cell Cycle and Replication, Phospho Specific
Primary
2322
Human, Mouse (Negative)