Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-Flt-3/Flk-2/CD135, FITC, Clone: BV10, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP154414

Catalog No. NBP154414


Add to cart

Description

Description

Flt-3/Flk-2/CD135 Monoclonal antibody specifically detects Flt-3/Flk-2/CD135 in Human samples. It is validated for Flow Cytometry.
Specifications

Specifications

Flt-3/Flk-2/CD135
BV10
FITC
PBS and 1.0% BSA with 0.09% Sodium Azide
CD135, CD135 antigen, EC 2.7.10, fetal liver kinase 2, FL cytokine receptor, FLK2, FLT-3, FLT3 receptor tyrosine kinase, Fms-like tyrosine kinase 3, fms-related tyrosine kinase 3, growth factor receptor tyrosine kinase type III, Stem cell tyrosine kinase 1, STK-1, STK1EC 2.7.10.1, Tyrosine-protein kinase receptor FLT3
Mouse
IgG1
1 ml
Cell Cycle and Replication
Primary
2322
Flow Cytometry
0.5 mg/mL
Flow Cytometry 1:10-1:1000
Purified
FLT3
This hybridoma was established by fusion of mouse myeloma cell SP2/0 with Balb/c mouse splenocyte immunized with the pro B-cell line, BV-173.
Protein A purified
RUO
Store at 4C in the dark.
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only