Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-Flt-3/Flk-2/CD135, PerCP, Clone: BV10A4, Novus Biologicals™

Manufacturer:  Novus Biologicals NBP142792PCP

Catalog No. NBP142792P

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Specifications

Specifications

Flt-3/Flk-2/CD135
BV10A4
Flow Cytometry, Immunoprecipitation
Purified
FLT3
BV-173 leukemic cell line
Protein A purified
RUO
Store at 4C in the dark.
Monoclonal
The mouse monoclonal antibody BV10A4 (BV10) reacts with CD135 (FLT3, FLK2, STK-1), a 130-160 kDa type III receptor tyrosine kinase that is involved in early steps of hematopoiesis.
Flow Cytometry, Immunoprecipitation
PerCP
PBS with 0.05% Sodium Azide
CD135, CD135 antigen, EC 2.7.10, fetal liver kinase 2, FL cytokine receptor, FLK2, FLT-3, FLT3 receptor tyrosine kinase, Fms-like tyrosine kinase 3, fms-related tyrosine kinase 3, growth factor receptor tyrosine kinase type III, Stem cell tyrosine kinase 1, STK-1, STK1EC 2.7.10.1, Tyrosine-protein kinase receptor FLT3
Mouse
IgG1
0.1 ml
Cell Cycle and Replication, Phospho Specific
Primary
2322
Human, Mouse (Negative)
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only