Learn More
anti-FOXO1A/3/4, DyLight 680, Clone: 14E4, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus Biologicals NBP141195FR
Catalog No. NBP141195FR
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
FOXO1A/3/4 Monoclonal antibody specifically detects FOXO1A/3/4 in Human samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.
Specifications
Specifications
FOXO1A/3/4
140000
Western Blot, Immunocytochemistry/Immunofluorescence
Purified
FOXO1
Amino acids 242-655 of FKHR expressed in E. coli.
Protein G purified
RUO
Store at 4C in the dark.
Monoclonal
This antibody will recognize human FKHR.
Western Blot, Immunocytochemistry, Immunofluorescence
DyLight 680
50mM Sodium Borate with 0.05% Sodium Azide
FKH1, FKHRforkhead box protein O1, forkhead box O1, Forkhead box protein O1A, forkhead homolog in rhabdomyosarcoma, Forkhead in rhabdomyosarcoma, FOXO1Aforkhead, Drosophila, homolog of, in rhabdomyosarcoma
Mouse
IgG2b
0.1 ml
Apoptosis, Cancer, DNA Repair, Growth and Development, Neuronal Cell Markers
Primary
2308
Human