Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Frizzled-7 Goat anti-Mouse, Polyclonal, R&D Systems™
Goat Polyclonal Antibody has been used in 5 publications
Supplier: R&D Systems AF198
This item is not returnable.
View return policy
Description
Frizzled-7 Polyclonal specifically detects Frizzled-7 in Mouse samples. It is validated for Western Blot, Immunohistochemistry.Specifications
Frizzled-7 | |
Polyclonal | |
Unconjugated | |
Q61090 | |
FZD7 | |
Mouse myeloma cell line NS0-derived recombinant mouse Frizzled-7 Gln33-Leu185 Accession # Q61090 | |
100 μg | |
Primary | |
Detects mouse Frizzled-7 in direct ELISAs and Western blots. In direct ELISAs, approximately 15% cross-reactivity with recombinant mouse (rm) Frizzled-2 and less than 5% cross-reactivity with rmFrizzled-3, rmFrizzled-4, rhFrizzled-5, rmFrizzled-6, rmFrizzled-8, and rmFrizzled-9 is observed. | |
Mouse | |
IgG |
Western Blot, Immunohistochemistry | |
LYOPH | |
Western Blot 0.25 ug/mL, Immunohistochemistry 5-15 ug/mL | |
frizzled (Drosophila) homolog 7, frizzled homolog 7 (Drosophila), Frizzled, drosophila, homolog of, 7, Frizzled7, fz-7, FZD7, FzE3frizzled-7, hFz7 | |
Goat | |
Affinity Purified | |
RUO | |
8324 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title