Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-Frk, Clone: 1A8H2C12, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP128857

Catalog No. NBP128857


Add to cart

Description

Description

Frk Monoclonal antibody specifically detects Frk in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications

Specifications

Frk
1A8H2C12
Western Blot 1:500-1:2000, ELISA 1:10000, Immunohistochemistry, Immunohistochemistry-Paraffin
Ascites
FRK
Purified recombinant fragment of FRK(aa2-300) expressed in E. Coli.
Unpurified
RUO
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Monoclonal
Specific for FRK.
ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot
Unconjugated
Ascites with 0.03% Sodium Azide
EC 2.7.10, fyn-related kinaseProtein-tyrosine kinase 5, GTKEC 2.7.10.2, Nuclear tyrosine protein kinase RAK, PTK5 protein tyrosine kinase 5, RAKPTK5, tyrosine-protein kinase FRK
Mouse
IgG1
0.1 ml
Cell Cycle and Replication
Primary
2444
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only