Learn More
anti-FUBP3, Clone: 1216CT820.155.150, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP232296
Catalog No. NBP232296
Add to cart
Description
Description
FUBP3 Monoclonal antibody specifically detects FUBP3 in Human samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence.
Specifications
Specifications
FUBP3
1216CT820.155.150
Unconjugated
PBS with 0.09% Sodium Azide
Purified
FUBP3
Purified His-tagged FUBP3 protein was used to produced this monoclonal antibody.
61.64 kDa
400 ul
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Monoclonal
Human
Western Blot, ELISA, Immunocytochemistry, Immunofluorescence
0.5 mg/mL
Western Blot 1:2000, ELISA, Immunocytochemistry/Immunofluorescence 1:25
Q96I24
far upstream element (FUSE) binding protein 3, far upstream element-binding protein 3, FBP3, FLJ25229, FUSE-binding protein 3
Mouse
IgG1
Protein G purified
RUO
Primary
8939