Learn More
GABA-A R beta 1 Mouse anti-Human, Mouse, Rat, Alexa Fluor 647, Clone: S96-55, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus Biologicals NBP148319AF647
Catalog No. NBP148319Y
Add to cart
Description
Description
GABA-A R beta 1 Monoclonal antibody specifically detects GABA-A R beta 1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence.
Specifications
Specifications
GABA-A R beta 1
S96-55
Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence
P50571
GABA(A) receptor subunit beta-1, gamma-aminobutyric acid (GABA) A receptor, beta 1, gamma-aminobutyric acid receptor subunit beta-1
Mouse
IgG1
0.1 ml
GPCR, Neuronal Cell Markers, Neuroscience, Neurotransmission
Primary
2560
Human, Mouse, Rat
Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Western Blot
Alexa Fluor 647
50mM Sodium Borate with 0.05% Sodium Azide
Purified
GABRB1
Fusion protein amino acids 327-450 of mouse GABA-A-R-Beta 1
Protein G purified
RUO
Store at 4C in the dark.
Monoclonal
Detects approx 55kDa. No cross-reactivity against GABA-A-R-Beta 2 or -Beta3.