Learn More
anti-Galectin-3, Clone: B2C10, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus Biologicals NBP142384
Catalog No. NBP142384
Add to cart
Description
Description
Galectin-3 Monoclonal antibody specifically detects Galectin-3 in Human, Mouse samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Functional, Immunoassay.
Specifications
Specifications
Galectin-3
B2C10
Unconjugated
PBS, containing 0.1% bovine serum albumin. with No Preservative
Purified
LGALS3
The antibody reacts with the Galectin-3 antigen.
Protein A or G purified
RUO
Store at 4C. Do not freeze.
Monoclonal
The monoclonal antibody B2C10 reacts with galectin-3, a 30 kDa protein. Galectin-3 is a member of the galectin family. The protein is composed of three domains: a small amino-terminal domain, a carboxyl-terminal carbohydrate recognition domain (CRD) and amino-terminal domain containing repeating elements. Galectin-3 is normally distributed in epithelia of many organs and various inflammatory cells, including macrophages, as well as dendritic cells and Kupffer cells. The expression of this lectin is up-regulated during inflammation, cell proliferation, cell differentiation and through trans-activation by viral proteins. The expression is also affected by neoplastic transformation: up-regulated in certain types of lymphomas and thyroid carcinoma, while down-regulated in other types of malignancies, such as colon, breast, ovarian and uterine carcinomas. Galectin-3 has been shown to function through both intracellular and extracellular actions. Related to its intracellular functions, galectin-3 has been identified as a component of heterogeneous nuclear ribonuclear protein (hnRNP), a factor in pre-mRNA splicing, and has been found to control cell cycle and prevent T cell apoptosis. On the other hand, this protein has also been demonstrated to function as extracellular molecule in activating various types of cells, including monocytes/macrophages, mast cells, neutrophils and lymphocytes. Galectin-3 has been shown to mediate cell-cell and cell-extracellular matrix interactions. The monoclonal antibody B2C10 inhibits the binding of 125I-labeled galectin-3 to IgE coated on microtiter plates, the galecin-3's hemagglutination activity and galectin-3-induced superoxide production by human neutrophils. This inhibitory activity of B2C10 is probably the result of its disruption of the self-association process. The epitope of the monoclonal antibody B2C10 is found within the first 45 amino acids of galectin-3. The antibody B2C10 does not react with Galecin-3C and is cross reactive with mouse galectin-3.
Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot
0.1 mg/mL
Western Blot 1:100-1:2000, Flow Cytometry 1:10-1:1000, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:500, Immunohistochemistry-Frozen 1:10-1:500, Functional, Immunoassay
P17931
Carbohydrate-binding protein 35, CBP 35,35 kDa lectin, Gal-3, galactin-3, Galactose-specific lectin 3, Galactoside-binding protein, GALBPCBP35, galectin-3, GALIG, IgE-binding protein, L31, L-31, Laminin-binding protein, Lectin L-29, lectin, galactoside-binding, soluble, 3, LGALS2, MAC-2, Mac-2 antigen, MAC2GAL3
Mouse
IgG1
0.05 mg
Apoptosis, Cancer, Cellular Markers, Tumor Suppressors
Primary
3958
Human, Mouse