Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GAP-43 Mouse anti-Human, Mouse, Rat, Chicken, Rabbit, Reptile, Clone: GAP-8A12, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP141337

Catalog No. NBP141337

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

GAP-43 Monoclonal antibody specifically detects GAP-43 in Human, Mouse, Rat, Chicken, Rabbit, Reptile samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen).
Specifications

Specifications

GAP-43
GAP-8A12
Unconjugated
Lyophilized from 1.2% sodium acetate, with 2mg BSA and 0.01mg NaN3 as preservative. with 0.01 mg Sodium Azide
Purified
GAP43
Human plasma gelsolin.
Immunogen affinity purified
RUO
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Monoclonal
Human, Mouse, Rat, Chicken, Snake. May react with Rabbit.
Chicken, Human, Mouse, Rabbit, Rat, Reptile
Immunohistochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot
LYOPH
Western Blot 0.5-1 ug/ml, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 1-2 ug/ml, Immunohistochemistry-Frozen 1-2 ug/ml
P07936
Axonal membrane protein GAP-43, B-50, calmodulin-binding protein P-57, growth associated protein 43, Growth-associated protein 43, nerve growth-related peptide GAP43, Neural phosphoprotein B-50, neuromodulin, neuron growth-associated protein 43, PP46, protein F1
Mouse
IgG2a
0.1 mg
Cancer, Cellular Markers, Growth and Development, Neuronal Cell Markers, Neuroscience
Primary
2596
Reconstitute with deionized water.
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only