Learn More
anti-GAP-43, DyLight 488, Clone: 1E3, Novus Biologicals™
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP192713G
Catalog No. NBP192713G
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Specifications
Specifications
GAP-43
1000
Western Blot, Immunocytochemistry/Immunofluorescence
Purified
GAP43
C-terminal peptide of rat and mouse GAP43 (KEDPEADQEHA) to which an N-terminal Cysteine residue was added to allow chemical coupling to Keyhole Limpet Hemocyanin carrier protein.
43 kDa
0.1 ml
Cancer, Cellular Markers, Growth and Development, Neuronal Cell Markers, Neuroscience, Phospho Specific
Primary
2596
Immunocytochemistry, Immunofluorescence, Western Blot
DyLight 488
50mM Sodium Borate with 0.05% Sodium Azide
Axonal membrane protein GAP-43, B-50, calmodulin-binding protein P-57, growth associated protein 43, Growth-associated protein 43, nerve growth-related peptide GAP43, Neural phosphoprotein B-50, neuromodulin, neuron growth-associated protein 43, PP46, protein F1
Mouse
IgG1 Kappa
Protein G purified
RUO
Store at 4C in the dark.
Monoclonal
Human, Mouse, Rat