Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GDF-15 Mouse anti-Human, Clone: 23B3.D2.H5, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP142719

Catalog No. NBP142719


Add to cart

Description

Description

GDF-15 Monoclonal antibody specifically detects GDF-15 in Human samples. It is validated for Western Blot, ELISA.
Specifications

Specifications

GDF-15
23B3.D2.H5
Unconjugated
0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2 with 0.01% Sodium Azide
Purified
GDF15
This Protein A purified antibody was prepared by repeated immunizations with a synthetic peptide corresponding to a region near the carboxy terminal end of human GDF-15 protein. A residue of cysteine was added to facilitate coupling to KLH.
34.14 kDa
0.1 mg
Hepatic Stem Cell Markers, Stem Cell Markers
Primary
9518
Human, Monkey, Primate
ELISA, Western Blot
1.13 mg/mL
Western Blot 1:1000, ELISA 1:2000
Q99988
growth differentiation factor 15, growth/differentiation factor 15, Macrophage inhibitory cytokine 1, MIC-1NSAID-activated gene 1 protein, MIC1Prostate differentiation factor, NAG-1NSAID-regulated gene 1 protein, NSAID (nonsteroidal inflammatory drug)-activated protein 1, PDFGDF-15, PLABNRG-1, Placental bone morphogenetic protein, Placental TGF-beta, PTGFBPTGF-beta
Mouse
IgG1 Kappa
Protein A purified
RUO
Store at -20°C. Avoid freeze-thaw cycles.
Monoclonal
Partial reactivity is expected against rat based on an 86% identity with the immunizing sequence. Cross-reactivity with NAG-1 from other sources has not been determined.
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only