Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-GDF-8/Myostatin, Clone: 6H12, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP147388

Catalog No. NBP147388


Add to cart

Description

Description

GDF-8/Myostatin Monoclonal antibody specifically detects GDF-8/Myostatin in Human samples. It is validated for Western Blot, ELISA.
Specifications

Specifications

GDF-8/Myostatin
6H12
Western Blot 1: 500-1:2000, ELISA 1:10000
Ascites
MSTN
Purified recombinant fragment of GDF8/Myostatin expressed in E. coli.
42.7 kDa
0.1 ml
Cytokine Research
Primary
2660
ELISA, Western Blot
Unconjugated
Ascites with 0.03% Sodium Azide
GDF-8, GDF8growth differentiation factor 8, growth/differentiation factor 8, Myostatin
Mouse
IgG1
Unpurified
RUO
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only