Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-GFAP, Clone: SB61b, Novus Biologicals™

Manufacturer:  Novus Biologicals NBP128623

Catalog No. NBP128623

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Specifications

Specifications

GFAP
SB61b
Unconjugated
0.1M BBS (pH 8.0) with No Preservative
Purified
GFAP
Recombinant human Glial Fibrillary Acid Protein (GFAP)
50 kDa
0.5 mg
Astrocyte Markers, Cancer, Cytoskeleton Markers, Neuronal Stem Cell Markers, Neuroscience, Stem Cell Markers
Primary
2670
Human
Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Frozen)
0.5 mg/mL
Western Blot 2 ug/ml, ELISA 1:100-1:2000, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Frozen 5 ug/ml
P14136
FLJ45472, GFAP astrocytes, glial fibrillary acidic protein
Mouse
IgG2b Kappa
Protein A or G purified
RUO
Store at 4C. Do not freeze.
Monoclonal
Reacts with human GFAP (approx. 50 kDa)
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only