Learn More
GKAP/DLGAP1 Mouse anti-Human, Mouse, Rat, Alexa Fluor 700, Clone: S127-31, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus Biologicals NBP147612AF700
Catalog No. NBP147612Z
Add to cart
Description
Description
GKAP/DLGAP1 Monoclonal antibody specifically detects GKAP/DLGAP1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence.
Specifications
Specifications
GKAP/DLGAP1
S127-31
Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence
O14490
DAP1, DAP-1-ALPHA, DAP-1DAP-1-BETA, discs, large (Drosophila) homolog-associated protein 1, disks large-associated protein 1, GKAPMGC88156, hGKAP, PSD-95/SAP90 binding protein 1, PSD-95/SAP90-binding protein 1, SAP90/PSD-95-associated protein 1, SAPAP1Guanylate kinase-associated protein
Mouse
IgG2b
Protein G purified
RUO
Primary
9229
Human, Mouse, Rat
Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Western Blot
Alexa Fluor 700
50mM Sodium Borate with 0.05% Sodium Azide
Purified
DLGAP1
Fusion protein amino acids 772-992 (C-terminus) of rat SAPAP1
108 kDa
0.1 ml
Store at 4C in the dark.
Monoclonal
Detects approx. 120kDa (SAPAP2), approx. 110kDa (SAPAP1/3/4). It does cross-react with other SAPAPs .