Learn More
anti-GLP1, Alexa Fluor 488, Clone: HGL-B5, Novus Biologicals™
Manufacturer:  Novus Biologicals NBP150697AF488
Catalog No. NBP150697X
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Specifications
Specifications
GLP1
HGL-B5
ELISA, Immunocytochemistry/Immunofluorescence
P01275
glicentin-related polypeptide, GLP1, GLP2, glucagon, glucagon-like peptide 1, glucagon-like peptide 2, GRPP
Mouse
IgG1
0.1 ml
Cancer, GPCR, Signal Transduction
Primary
2641
ELISA, Immunocytochemistry, Immunofluorescence
Alexa Fluor 488
50mM Sodium Borate with 0.05% Sodium Azide
Affinity Purified
GCG
Full length synthetic human GLP-1 (31 amino acids)
Protein A or G purified
RUO
Store at 4C in the dark.
Monoclonal
Human