Learn More
anti-Glyoxalase I, Alexa Fluor 488, Clone: Glo1a, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus Biologicals NBP119015AF488
Catalog No. NBP119015X
Add to cart
Description
Description
Glyoxalase I Monoclonal antibody specifically detects Glyoxalase I in Human, Mouse samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence.
Specifications
Specifications
Glyoxalase I
Glo1a
Western Blot, ELISA, Immunocytochemistry/Immunofluorescence
Q04760
Aldoketomutase, EC 4.4.1.5, GLOD1, Glx I, GLYI, glyoxalase domain containing 1, glyoxalase Ialdoketomutase, Ketone-aldehyde mutase, lactoyl glutathione lyase, lactoylglutathione lyase, methylglyoxalase, S-D-lactoylglutathione methylglyoxal lyase
Mouse
IgG1 Kappa
Protein G purified
RUO
Store at 4C in the dark.
Monoclonal
Human, Mouse
ELISA, Immunocytochemistry, Immunofluorescence, Western Blot
Alexa Fluor 488
50mM Sodium Borate with 0.05% Sodium Azide
Purified
GLO1
Full length human GLO1 protein [Swiss-Prot #Q04760].
21 kDa
0.1 ml
Cancer, Core ESC Like Genes, Signal Transduction, Stem Cell Markers
Primary
2739