Learn More
anti-Golgi Complex, DyLight 680, Clone: SPM581, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP234813FR
Catalog No. NBP234813FR
Add to cart
Description
Description
Golgi Complex Monoclonal antibody specifically detects Golgi Complex in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.
Specifications
Specifications
Golgi Complex
SPM581
Immunocytochemistry/Immunofluorescence
Purified
SU-DHL-1 large cell lymphoma cells.
Protein A purified
RUO
Primary
This MAb recognizes Golgi complex in human cells. It is a part of a new panel of reagents, which recognizes subcellular organelles or compartments of human cells. These markers may be useful in identification of these organelles in cells, tissues, and biochemical preparations. It recognizes an antigen associated with the Golgi complex in human cells only. It can be used to stain the Golgi complex in cell or tissue preparations and can be used as a Golgi marker in subcellular fractions. It produces a diffuse staining pattern of the Golgi zone in normal and malignant cells and may be used to stain Golgi complex of cells in frozen tissue sections. It can also be used with paraformaldehyde fixed frozen tissue or cell preparations. This MAb is an excellent marker for human cells in xenographic model research. It reacts specifically with human cells.
Immunocytochemistry, Immunofluorescence
DyLight 680
50mM Sodium Borate with 0.05% Sodium Azide
Mouse
IgG1 Kappa
0.1 ml
Store at 4C in the dark.
Monoclonal
Human