Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-Goosecoid, Clone: 1D7, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP147790

Catalog No. NBP147790


Add to cart

Description

Description

Goosecoid Monoclonal antibody specifically detects Goosecoid in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications

Specifications

Goosecoid
OTI1D7
Unconjugated
PBS (pH 7.3), 1.0% BSA and 50% Glycerol with 0.02% Sodium Azide
goosecoid homeobox, homeobox protein goosecoid
Mouse
IgG1
Affinity purified
RUO
Primary
145258
Human
Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot
0.7 mg/mL
Western Blot 1:500-1000, Flow Cytometry 1:100, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 1:50
Affinity Purified
GSC
Human recombinant protein fragment corresponding to amino acids 107-257 of human GSC (NP_776248) produced in E.coli.
28 kDa
0.1 ml
Store at -20°C. Avoid freeze-thaw cycles.
Monoclonal
This antibody is specific for Homo sapiens goosecoid homeobox (GSC).
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only