Learn More
anti-Granulocytes, HRP, Clone: BM-2, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP233114H
Catalog No. NBP233114H
Add to cart
Description
Description
Myeloid Cell Marker Monoclonal antibody specifically detects Myeloid Cell Marker in Human, Primate samples. It is validated for ELISA, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.
Specifications
Specifications
Granulocytes
BM-2
ELISA, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen
Purified
CSF3
Nuclei from pokeweed mitogen stimulated human peripheral blood lymphocytes were used as the immunogen for this granulocyte antibody.
Protein G purified
RUO
Primary
1440
Human, Primate
ELISA, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen)
HRP
PBS with No Preservative
C17orf33, chromosome 17 open reading frame 33, colony stimulating factor 3 (granulocyte), CSF3OS, filgrastim, G-CSF, GCSFlenograstim, granulocyte colony-stimulating factor, Lenograstim, MGC45931, pluripoietin
Mouse
IgG1
0.1 ml
Store at 4C in the dark.
Monoclonal
Recognizes an unidentified antigen in the cytoplasm of mature granulocytes. It shows no reactivity with any other cell type in human tissues. Markers of myeloid cells are useful in the identification of different levels of cellular differentiation. MAb BM-2 reacts with early precursor and mature forms of human myeloid cells. This MAb is useful for the detection of myeloid leukemias and granulocytic sarcomas. It can be used as a marker of granulocytes in normal tissues or inflammatory processes.