Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-Growth Hormone, Clone: 8J33, Novus Biologicals™

Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP236217

Catalog No. NBP236217

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Specifications

Specifications

Growth Hormone
8J33
Unconjugated
PBS with No Preservative
GHIGHD1B, GHNGrowth hormone, GH-Nsomatotropin, growth hormone 1Pituitary growth hormone, hGH-N
Mouse
IgG2
0.1 mg
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Monoclonal
Reconstitute with 0.2 ml sterilized PBS to a final concentration of 0.5 mg/ml.
Blocking Assay, Western Blot
LYOPH
Western Blot 1:500 - 1:1000, Blocking/Neutralizing 50 - 200 molar excess
Purified
GH1
recombinant human Growth Hormone
Protein G purified
RUO
Primary
2688
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only