Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-GSK-3 beta, HRP, Clone: 3A11, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP128587

Catalog No. NBP128587


Add to cart

Description

Description

GSK-3 beta Monoclonal antibody specifically detects GSK-3 beta in Human samples. It is validated for Western Blot, ELISA.
Specifications

Specifications

GSK-3 beta
3A11
Western Blot 1:100-1:2000, ELISA 1:1000-1:2000
Purified
GSK3B
The immunogen is a peptide made to the amino acid region Human GSK-3 beta
Protein A or G purified
RUO
Store at 4C. Do not freeze.
Monoclonal
Human GSK-3 beta
ELISA, Western Blot
HRP
Stock solution in 50% Glycerol/50% PBS (pH 7.4) with No Preservative
EC 2.7.11, EC 2.7.11.26, glycogen synthase kinase 3 beta, glycogen synthase kinase-3 beta, GSK-3 beta, GSK3beta isoform
Mouse
IgG2a
0.5 ml
Wnt Signaling Pathway
Primary
2932
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only